CAS RN 39516-03-5
IUPAC Name:
4-phenylbutan-2-ol
Synonyms:
2-hydroxy-4-phenylbutane
Molecular Weight (g/mol):
150.22
Molecular Formula:
C10H14O
InChi Key:
GDWRKZLROIFUML-UHFFFAOYNA-N
SMILES:
CC(O)CCC1=CC=CC=C1
1
–
2
of
2
results
1
Sigma Aldrich (S)-(+)-4-Phenyl-2-Butanol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.