CAS RN 39565-05-4
IUPAC Name:
5-(4-nitrobenzenesulfonyl)-1,3-thiazol-2-amine
Synonyms:
5-(4-nitrobenzenesulfonyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol):
285.29
Molecular Formula:
C9H7N3O4S2
InChi Key:
GKTKCGAOXFHFTD-UHFFFAOYSA-N
SMILES:
NC1=NC=C(S1)S(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
2-Amino-5-(4-nitrophenylsulfonyl)-thiazole, Thermo Scientific™
CAS: 39565-05-4 Molecular Formula: C9H7N3O4S2 Molecular Weight (g/mol): 285.292 InChI Key: GKTKCGAOXFHFTD-UHFFFAOYSA-N Synonym: 2-amino-5-4-nitrophenylsulfonyl thiazole,2-amino-5-p-nitrophenylsulfonyl thiazole,2-amino-5-p-nitrobenzenesulfonyl thiazole,5-4-nitrophenyl sulfonyl-1,3-thiazol-2-amine,2-thiazolamine, 5-4-nitrophenyl sulfonyl,thiazole, 2-amino-5-p-nitrophenyl sulfonyl,thiazole, 2-amino-5-p-nitrophenylsulfonyl,p-nitrophenyl-2-aminothiazol-5-ylsulphone,wln: t5n csj bz dswr dnw PubChem CID: 1535 IUPAC Name: 5-(4-nitrophenyl)sulfonyl-1,3-thiazol-2-amine SMILES: C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)C2=CN=C(S2)N