CAS RN 3958-82-5
IUPAC Name:
2-bromocyclohexa-2,5-diene-1,4-dione
Synonyms:
2-bromocyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol):
186.99
Molecular Formula:
C6H3BrO2
InChi Key:
PYEQXZOIQJWYAH-UHFFFAOYSA-N
SMILES:
BrC1=CC(=O)C=CC1=O
1
–
1
of
1
results
1
2-Bromo-1,4-benzoquinone 93.0+%, TCI America™
CAS: 3958-82-5 Molecular Formula: C6H3BrO2 Molecular Weight (g/mol): 186.992 MDL Number: MFCD00189377 InChI Key: PYEQXZOIQJWYAH-UHFFFAOYSA-N Synonym: 2-Bromocyclohexadiene-1,4-dione, Bromoquinone PubChem CID: 151181 IUPAC Name: 2-bromocyclohexa-2,5-diene-1,4-dione SMILES: C1=CC(=O)C(=CC1=O)Br