accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOOSOOSHOOO

CAS RN 39616-93-8

IUPAC Name: 3-(3-sulfobenzenesulfonyl)benzene-1-sulfonic acid
Synonyms: 3-(3-sulfobenzenesulfonyl)benzenesulfonic acid
Molecular Weight (g/mol): 378.38
Molecular Formula: C12H10O8S3
InChi Key: FEOSEXDRZIIBNX-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC(=CC=C1)S(=O)(=O)C1=CC(=CC=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results