accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHO

CAS RN 39627-84-4

IUPAC Name: naphthalene-2-carbohydrazide
Synonyms: naphthalene-2-carbohydrazide
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: RARLPRMZJNIQGU-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)