accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONH2OH

CAS RN 39748-77-1

IUPAC Name: 2-amino-3-(4-hydroxyphenyl)propanoic acid
Synonyms: tyrosin tirosina
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: OUYCCCASQSFEME-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(O)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results