accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3O2H

CAS RN 3979-51-9

IUPAC Name: propan-2-(²H)ol
Synonyms: propan-2-(²H)ol
Molecular Weight (g/mol): 61.1
Molecular Formula: C3H8O
InChi Key: KFZMGEQAYNKOFK-QYKNYGDISA-N
SMILES: [2H]OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results