CAS RN 3987-53-9
IUPAC Name:
2-[benzyl(carboxymethyl)amino]acetic acid
Synonyms:
[benzyl(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol):
223.23
Molecular Formula:
C11H13NO4
InChi Key:
SZQUPQVVCLFZLC-UHFFFAOYSA-N
SMILES:
OC(=O)CN(CC(O)=O)CC1=CC=CC=C1
1
–
1
of
1
results
1
N-Benzyliminodiacetic Acid 98.0+%, TCI America™
CAS: 3987-53-9 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.228 MDL Number: MFCD00004285 InChI Key: SZQUPQVVCLFZLC-UHFFFAOYSA-N PubChem CID: 77606 IUPAC Name: 2-[benzyl(carboxymethyl)amino]acetic acid SMILES: C1=CC=C(C=C1)CN(CC(=O)O)CC(=O)O