CAS RN 39905-50-5
IUPAC Name:
4-(pentyloxy)aniline
Synonyms:
4-(pentyloxy)aniline
Molecular Weight (g/mol):
179.26
Molecular Formula:
C11H17NO
InChi Key:
QZLNSNIHXKQIIS-UHFFFAOYSA-N
SMILES:
CCCCCOC1=CC=C(N)C=C1
1
–
1
of
1
results
1
4-Pentyloxyaniline, 99%, Thermo Scientific™
CAS: 39905-50-5 Molecular Formula: C11H17NO Molecular Weight (g/mol): 179.26 MDL Number: MFCD00007867 InChI Key: QZLNSNIHXKQIIS-UHFFFAOYSA-N Synonym: 4-pentyloxyaniline,p-pentyloxyaniline,4-pentyloxy aniline,benzenamine, 4-pentyloxy,4-n-pentoxyaniline,4-pentyloxyphenylamine,p-pentyloxy aniline,p-n-pentoxy aniline,4-pentyloxy benzenamine,p-aminophenyl pentyl ether PubChem CID: 94500 IUPAC Name: 4-(pentyloxy)aniline SMILES: CCCCCOC1=CC=C(N)C=C1