accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3993-78-0

IUPAC Name: 4-chloropyrimidin-2-amine
Synonyms: 4-chloropyrimidin-2-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: DBGFGNCFYUNXLD-UHFFFAOYSA-N
SMILES: NC1=NC=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)