Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chloropyrimidine, 95%, Thermo Scientific™

Catalog No. AC454060010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454060010 Supplier Thermo Scientific Chemicals Supplier No. 454060010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3993-78-0
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
InChI Key DBGFGNCFYUNXLD-UHFFFAOYSA-N
Synonym 2-amino-4-chloropyrimidine, 2-amine-4-chloropyrimidine, 2-pyrimidinamine, 4-chloro, 4-chloro-2-pyrimidinamine, 4-chloro-2-aminopyrimidine, 4-chloropyrimidine-2-amine, 4-chloro-pyrimidin-2-ylamine, 4-chloropyrimidine-2-ylamine, 2-amino-6-chloropyrimidine, 4-chloro-2-pyrimidineamine
PubChem CID 223332
IUPAC Name 4-chloropyrimidin-2-amine
SMILES C1=CN=C(N=C1Cl)N

Specifications

Melting Point 155°C to 160°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 129.55
Percent Purity 95%
Chemical Name or Material 2-Amino-4-chloropyrimidine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.