accessibility menu, dialog, popup

UserName

Viewing profile as user:
SBr

CAS RN 40133-22-0

IUPAC Name: 2-(4-bromophenyl)thiophene
Synonyms: 2-(4-bromophenyl)thiophene
Molecular Weight (g/mol): 239.13
Molecular Formula: C10H7BrS
InChi Key: XKOJJKOSNQXQGP-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results