CAS RN 402713-81-9
IUPAC Name:
N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-(piperazin-1-yl)benzene-1-sulfonamide hydrochloride
Synonyms:
N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-(piperazin-1-yl)benzenesulfonamide hydrochloride
Molecular Weight (g/mol):
482.8
Molecular Formula:
C18H22Cl3N3O4S
InChi Key:
RNKCEBCFUSXSQE-UHFFFAOYSA-N
SMILES:
Cl.COC1=C(C=C(C=C1)S(=O)(=O)NC1=CC(Cl)=CC(Cl)=C1OC)N1CCNCC1
1
–
1
of
1
results
1
R&D Systems™ SB 399885 hydrochloride
CAS: 402713-81-9 Molecular Formula: C18H22Cl3N3O4S