accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFCH3NNH3CH3C

CAS RN 402846-78-0

IUPAC Name: 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 240.05
Molecular Formula: C9H17BF4N2
InChi Key: VCAIYEJBOWHUGP-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)