CAS RN 40297-09-4
IUPAC Name:
N-(2-phenylcyclopentyl)-1-azacyclotridec-1-en-2-amine hydrochloride
Synonyms:
N-(2-phenylcyclopentyl)-1-azacyclotridec-1-en-2-amine hydrochloride
Molecular Weight (g/mol):
377.01
Molecular Formula:
C23H37ClN2
InChi Key:
CKOPQUCSDBVAQG-UHFFFAOYNA-N
SMILES:
Cl.C1CC(NC2=NCCCCCCCCCCC2)C(C1)C1=CC=CC=C1
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MDL-12,330A, Hydrochloride
Cell-permeable and irreversible inhibitor of adenylate cyclase (IC50 = 250 μM).