accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SHOOOHO

CAS RN 4033-46-9

IUPAC Name: 2-amino-3-[(2-carboxyethyl)sulfanyl]propanoic acid
Synonyms: 2-amino-3-[(2-carboxyethyl)sulfanyl]propanoic acid
Molecular Weight (g/mol): 193.22
Molecular Formula: C6H11NO4S
InChi Key: FBPINGSGHKXIQA-UHFFFAOYNA-N
SMILES: NC(CSCCC(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results