accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 403842-38-6

IUPAC Name: 2-[2-(hex-1-yn-1-yl)-6-(methylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-[2-(hex-1-yn-1-yl)-6-(methylamino)purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 361.4
Molecular Formula: C17H23N5O4
InChi Key: KOCIMZNSNPOGOP-UHFFFAOYNA-N
SMILES: CCCCC#CC1=NC(NC)=C2N=CN(C3OC(CO)C(O)C3O)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results