CAS RN 40447-21-0
IUPAC Name:
5-[(propan-2-yl)amino]pentan-1-ol
Synonyms:
5-(isopropylamino)pentan-1-ol
Molecular Weight (g/mol):
145.25
Molecular Formula:
C8H19NO
InChi Key:
UEXRIBOEINFZJS-UHFFFAOYSA-N
SMILES:
CC(C)NCCCCCO
1
–
1
of
1
results
1
5-(Isopropylamino)pentanol 98.0+%, TCI America™
CAS: 40447-21-0 Molecular Formula: C8H19NO Molecular Weight (g/mol): 145.246 InChI Key: UEXRIBOEINFZJS-UHFFFAOYSA-N PubChem CID: 414092 IUPAC Name: 5-(propan-2-ylamino)pentan-1-ol SMILES: CC(C)NCCCCCO