accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3

CAS RN 40458-98-8

IUPAC Name: 2,7-bis(propan-2-yl)naphthalene
Synonyms: 2,7-diisopropylnaphthalene
Molecular Weight (g/mol): 212.34
Molecular Formula: C16H20
InChi Key: YGDMAJYAQCDTNG-UHFFFAOYSA-N
SMILES: CC(C)C1=CC2=CC(=CC=C2C=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results