accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)(R)H2N

CAS RN 40571-86-6

IUPAC Name: (1R,2R)-N-benzyl-2-hydroxycyclohexan-1-aminium
Synonyms: (1R,2R)-N-benzyl-2-hydroxycyclohexan-1-aminium
Molecular Weight (g/mol): 206.31
Molecular Formula: C13H20NO
InChi Key: NJNCYFUGUYIMEQ-CHWSQXEVSA-O
SMILES: O[C@@H]1CCCC[C@H]1[NH2+]CC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results