Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Benzylamino-1-cyclohexanol, 99+%, Thermo Scientific™

Catalog No. AC265840050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC265840050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC265840050 Supplier Thermo Scientific Chemicals Supplier No. 265840050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40571-86-6
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD00145421
InChI Key NJNCYFUGUYIMEQ-CHWSQXEVSA-N
Synonym 1r,2r-2-benzylamino cyclohexanol, trans-2-benzylamino cyclohexanol, trans-2-benzylamino-1-cyclohexanol, 1r,2r-2-benzylamino cyclohexan-1-ol, 1r,2r-2-benzylamino-1-cyclohexanol, cyclohexanol, 2-phenylmethyl amino-, 1r-trans, 2-benzylamino-cyclohexanol, trans-2-benzylamino-cyclohexanol, pubchem19547, trans-2-benzylaminocyclohexan-1-ol
PubChem CID 2724650
IUPAC Name (1R,2R)-2-(benzylamino)cyclohexan-1-ol
SMILES C1CCC(C(C1)NCC2=CC=CC=C2)O

Specifications

Melting Point 69°C to 72°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 5 g
Formula Weight 205.3
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Benzylamino-1-cyclohexanol, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.