CAS RN 405911-17-3
IUPAC Name:
2-{3-[3-({[2-chloro-3-(trifluoromethyl)phenyl]methyl}(2,2-diphenylethyl)amino)propoxy]phenyl}acetic acid hydrochloride
Synonyms:
C33H31ClF3NO3 hydrochloride
Molecular Weight (g/mol):
618.52
Molecular Formula:
C33H32Cl2F3NO3
InChi Key:
NMPUWJFHNOUNQU-UHFFFAOYSA-N
SMILES:
Cl.OC(=O)CC1=CC(OCCCN(CC(C2=CC=CC=C2)C2=CC=CC=C2)CC2=C(Cl)C(=CC=C2)C(F)(F)F)=CC=C1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific™ GW3965 hydrochloride, 95%
Thermo Scientific™ GW3965 hydrochloride, 95%
2
R&D Systems™ GW 3965 hydrochloride
CAS: 405911-17-3 Molecular Formula: C33H32Cl2F3NO3