CAS RN 40600-13-3
IUPAC Name:
2-[(2-cyclopropylphenoxy)methyl]-4,5-dihydro-1H-imidazol-3-ium chloride
Synonyms:
cirazoline
Molecular Weight (g/mol):
252.74
Molecular Formula:
C13H17ClN2O
InChi Key:
XFRXWLWUUDJHPB-UHFFFAOYSA-N
SMILES:
[Cl-].C(OC1=CC=CC=C1C1CC1)C1=[NH+]CCN1
1
–
1
of
1
results
1
R&D Systems™ Cirazoline hydrochloride
CAS: 40600-13-3 Molecular Formula: C13H17ClN2O