accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNHClBr

CAS RN 40616-75-9

IUPAC Name: [(2-bromophenyl)methyl](2-chloroethyl)ethylazanium chloride
Synonyms: [(2-bromophenyl)methyl](2-chloroethyl)ethylazanium chloride
Molecular Weight (g/mol): 313.06
Molecular Formula: C11H16BrCl2N
InChi Key: NDDRNRRNYOULND-UHFFFAOYNA-N
SMILES: [Cl-].CC[NH+](CCCl)CC1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)