CAS RN 40732-48-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphate, Sodium Salt
Important intracellular second messenger implicated in the action of nitric oxide in many cell types.