accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNH3CCH3CH3

CAS RN 40739-43-3

IUPAC Name: trioctyl(propyl)azanium chloride
Synonyms: trioctyl(propyl)azanium chloride
Molecular Weight (g/mol): 432.22
Molecular Formula: C27H58ClN
InChi Key: WFEXFNMTEBFLMM-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCC[N+](CCC)(CCCCCCCC)CCCCCCCC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results