CAS RN 407587-33-1
IUPAC Name:
6-tert-butyl-2-(furan-2-amido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid
Synonyms:
6-tert-butyl-2-(furan-2-amido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylic acid
Molecular Weight (g/mol):
347.43
Molecular Formula:
C18H21NO4S
InChi Key:
ACLUEOBQFRYTQS-UHFFFAOYNA-N
SMILES:
CC(C)(C)C1CCC2=C(C1)SC(NC(=O)C1=CC=CO1)=C2C(O)=O