accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HN(R)(S)OOClCl

CAS RN 40796-97-2

IUPAC Name: (1R,5S)-3-(3,5-dichlorobenzoyloxy)-8-methyl-8-azabicyclo[3.2.1]octan-8-ium
Synonyms: bemesetron
Molecular Weight (g/mol): 315.21
Molecular Formula: C15H18Cl2NO2
InChi Key: MNJNPLVXBISNSX-PBWFPOADSA-O
SMILES: C[NH+]1[C@H]2CC[C@@H]1CC(C2)OC(=O)C1=CC(Cl)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)