accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)O

CAS RN 4087-39-2

IUPAC Name: (4aS)-4a-methyl-2,3,4,4a,5,6,7,8-octahydronaphthalen-2-one
Synonyms: (4aS)-4a-methyl-3,4,5,6,7,8-hexahydronaphthalen-2-one
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: OHERZLWVBJCXOF-NSHDSACASA-N
SMILES: C[C@@]12CCCCC1=CC(=O)CC2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results