accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40876-98-0

IUPAC Name: sodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Synonyms: sodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Molecular Weight (g/mol): 210.16
Molecular Formula: C8H11NaO5
InChi Key: JPTKZRPOIUYFTM-UHFFFAOYSA-N
SMILES: [Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)