Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl oxalacetate sodium salt, 95%, pract.

Catalog No. AC407375000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC407375000 500 g
AC407371000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC407375000 Supplier Thermo Scientific Chemicals Supplier No. 407375000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 40876-98-0 | C8H11NaO5 | 210.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40876-98-0
Molecular Formula C8H11NaO5
Molecular Weight (g/mol) 210.16
MDL Number MFCD00035571
InChI Key UJZUICGIJODKOS-UHFFFAOYSA-M
Synonym diethyl oxalacetate sodium salt, sodium z-1,4-diethoxy-1,4-dioxo-but-2-en-2-olate, ethyl sodium oxaloacetate, sodium-1,4-diethoxy-1,4-dioxobut-2-en-2-olate, ksc915o5n
PubChem CID 44134875
IUPAC Name sodium;1,4-diethoxy-1,4-dioxobut-2-en-2-olate
SMILES CCOC(=O)C=C(C(=O)OCC)[O-].[Na+]

Specifications

Color Beige to Orange
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Linear Formula C2H5OCOCH=C(ONa)COOC2H5
Quantity 500 g
Merck Index 14, 3836
Formula Weight 210.16
Percent Purity 95%
Grade Practical
Physical Form Crystalline Powder
Chemical Name or Material Diethyl oxalacetate sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 255-122-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.