accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHNOClOHO

CAS RN 4090-17-9

IUPAC Name: 2-(2-chloroacetamido)-3-methylbutanoic acid
Synonyms: 2-(2-chloroacetamido)-3-methylbutanoic acid
Molecular Weight (g/mol): 193.63
Molecular Formula: C7H12ClNO3
InChi Key: LJRISAYPKJORFZ-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)CCl)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results