accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NSN(R)O(R)OPOHOOPOHOO(R)O(R)(R)OH(S)OHNNNH2NN(S)OH(R)O

CAS RN 4090-29-3

IUPAC Name: 1-[(2R,3R,4S,5R)-5-[({[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)methyl]-4-hydroxy-3-oxidooxolan-2-yl]-3-carbamothioyl-1λ⁵-pyridin-1-ylium
Synonyms: 1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy(hydroxy)phosphoryl)oxy]methyl}-4-hydroxy-3-oxidooxolan-2-yl]-3-carbamothioyl-1λ⁵-pyridin-1-ylium
Molecular Weight (g/mol): 679.49
Molecular Formula: C21H27N7O13P2S
InChi Key: CXONXVMMINSQBV-NNYOXOHSSA-N
SMILES: NC(=S)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1[O-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results