accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3COClClHOO

CAS RN 4091-99-0

IUPAC Name: 2-[3,6-bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic acid
Synonyms: 2-[3,6-bis(acetyloxy)-2,7-dichloro-9H-xanthen-9-yl]benzoic acid
Molecular Weight (g/mol): 487.29
Molecular Formula: C24H16Cl2O7
InChi Key: PXEZTIWVRVSYOK-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC2=C(C=C1Cl)C(C1=CC(Cl)=C(OC(C)=O)C=C1O2)C1=CC=CC=C1C(O)=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results