accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONO

CAS RN 409345-29-5

IUPAC Name: 7-(4-methoxycyclohexanecarbonyl)-2H,3H,4H-pyrano[2,3-b]quinoline
Synonyms: 7-(4-methoxycyclohexanecarbonyl)-2H,3H,4H-pyrano[2,3-b]quinoline
Molecular Weight (g/mol): 325.41
Molecular Formula: C20H23NO3
InChi Key: QOTAQTRFJWLFCR-UHFFFAOYSA-N
SMILES: COC1CCC(CC1)C(=O)C1=CC=C2N=C3OCCCC3=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)