accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CSeSiCH3CH3H3C

CAS RN 4099-46-1

IUPAC Name: trimethyl[(trimethylsilyl)selanyl]silane
Synonyms: trimethyl[(trimethylsilyl)selanyl]silane
Molecular Weight (g/mol): 225.35
Molecular Formula: C6H18SeSi2
InChi Key: FKIZDWBGWFWWOV-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[Se][Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results