accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHSOHOO

CAS RN 4112-03-2

IUPAC Name: N-methylsulfamic acid
Synonyms: N-methylsulfamic acid
Molecular Weight (g/mol): 111.12
Molecular Formula: CH5NO3S
InChi Key: MYMDOKBFMTVEGE-UHFFFAOYSA-N
SMILES: CNS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results