accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4114-28-7

H3COONHHNOOCH3

CAS RN 4114-28-7

IUPAC Name: N'-(ethoxycarbonyl)ethoxycarbohydrazide
Synonyms: urethane (ethyl carbamate)
Molecular Weight (g/mol): 176.17
Molecular Formula: C6H12N2O4
InChi Key: JXMLAPZRDDWRRV-UHFFFAOYSA-N
SMILES: CCOC(=O)NNC(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results