accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 41175-50-2

IUPAC Name: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol
Synonyms: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: FOFUWJNBAQJABO-UHFFFAOYSA-N
SMILES: OC1=CC=C2CCCN3CCCC1=C23
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)