Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Hydroxyjulolidine, 97%

Catalog No. AAA1769206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1769206 5 g
AAA1769203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1769206 Supplier Thermo Scientific Chemicals Supplier No. A1769206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 41175-50-2 | C12H15NO | 189.258 g/mol

8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41175-50-2
Molecular Formula C12H15NO
MDL Number MFCD00006918
InChI Key FOFUWJNBAQJABO-UHFFFAOYSA-N
PubChem CID 170474

Specifications

Melting Point 132°C to 135°C
Quantity 5 g
Beilstein 1530423
Synonym 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
Solubility Information Soluble in water (partly),ethanol,chloroform,methanol,and acetone.
SMILES C1CC2=C3C(=C(C=C2)O)CCCN3C1
Molecular Weight (g/mol) 189.258
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 8-Hydroxyjulolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-247-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.