accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 41332-02-9

IUPAC Name: 1-(2-chloroethyl)naphthalene
Synonyms: 1-(2-chloroethyl)naphthalene
Molecular Weight (g/mol): 190.67
Molecular Formula: C12H11Cl
InChi Key: COIBFCUQUWJKEN-UHFFFAOYSA-N
SMILES: ClCCC1=CC=CC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results