accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNO

CAS RN 41361-28-8

IUPAC Name: hydrogen 1-ethylpiperidin-3-one chloride
Synonyms: hydrogen 1-ethylpiperidin-3-one chloride
Molecular Weight (g/mol): 163.65
Molecular Formula: C7H14ClNO
InChi Key: JLBJGYHSFYAUDJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN1CCCC(=O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results