accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41458-65-5

IUPAC Name: 2-amino-4,6-dimethylphenol
Synonyms: 2-amino-4,6-dimethylphenol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: GISWNAMJAQRJPC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(O)C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results