accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41498-71-9

IUPAC Name: bicyclo[2.2.1]heptan-2-yl formate
Synonyms: bicyclo[2.2.1]heptan-2-yl formate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: SGXIEZNAOCVSKO-UHFFFAOYNA-N
SMILES: O=COC1CC2CCC1C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results