accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(R)OH(S)OHNNHNNN

CAS RN 41552-82-3

IUPAC Name: (3R,4S,5R)-2-[6-(cyclopentylamino)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: N6-cyclopentyladenosine
Molecular Weight (g/mol): 335.36
Molecular Formula: C15H21N5O4
InChi Key: SQMWSBKSHWARHU-FJFSNTMWSA-N
SMILES: OC[C@H]1OC([C@H](O)[C@@H]1O)N1C=NC2=C(NC3CCCC3)N=CN=C12
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results