accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOO

CAS RN 4165-96-2

IUPAC Name: 3-phenylpentanedioate
Synonyms: 3-phenylpentanedioate
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: RZOKZOYSUCSPDF-UHFFFAOYSA-L
SMILES: [O-]C(=O)CC(CC([O-])=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results