accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNN

CAS RN 41814-78-2

IUPAC Name: 12-methyl-7-thia-2,4,5-triazatricyclo[6.4.0.0²,⁶]dodeca-1(12),3,5,8,10-pentaene
Synonyms: tricyclazole
Molecular Weight (g/mol): 189.24
Molecular Formula: C9H7N3S
InChi Key: DQJCHOQLCLEDLL-UHFFFAOYSA-N
SMILES: CC1=C2N3C=NN=C3SC2=CC=C1
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)