accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNH3

CAS RN 41838-46-4

IUPAC Name: 4-azaniumyl-1-methylpiperidin-1-ium
Synonyms: 4-ammonio-1-methylpiperidin-1-ium
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: ALOCUZOKRULSAA-UHFFFAOYSA-P
SMILES: C[NH+]1CCC([NH3+])CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results