accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNONNN

CAS RN 41864-22-6

IUPAC Name: 1-(1H-1,2,4-triazole-1-carbonyl)-1H-1,2,4-triazole
Synonyms: 1-(1,2,4-triazole-1-carbonyl)-1,2,4-triazole
Molecular Weight (g/mol): 164.13
Molecular Formula: C5H4N6O
InChi Key: YHNUDLCUIKMNSN-UHFFFAOYSA-N
SMILES: O=C(N1C=NC=N1)N1C=NC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)