Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Carbonyldi(1,2,4-triazole) 97.0+%, TCI America™
SDP

Catalog No. C23255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23255G 5 g
C232525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23255G Supplier TCI America Supplier No. C23255G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41864-22-6
Molecular Formula C5H4N6O
Molecular Weight (g/mol) 164.128
MDL Number MFCD00043399
InChI Key YHNUDLCUIKMNSN-UHFFFAOYSA-N
PubChem CID 4393961
IUPAC Name bis(1,2,4-triazol-1-yl)methanone
SMILES C1=NN(C=N1)C(=O)N2C=NC=N2

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
Synonym 1,1′C-Carbonylbis(1,2,4-triazole), CDT
Formula Weight 164.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Carbonyldi(1,2,4-triazole)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.